missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzanilide, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 148550010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Benzanilide | |
93-98-1 | |
100.0 | |
Authentic | |
Plastic bottle | |
C6H5CONHC6H5 | |
1kg | |
15, 1063 | |
Solubility in water: insoluble. Other solubilities: 1g/60 mL alcohol,1g/7 mL boiling alcohol,slightly soluble in ether,acetic acid | |
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2 | |
197.24 | |
197.24 |
98% | |
97.5 | |
117°C (10.0mmHg) | |
97.5% min. (HPLC) | |
C13H11NO | |
MFCD00003069 | |
12, 262 | |
benzanilide, n-benzoylaniline, benzamide, n-phenyl, n-phenyl-benzamide, benzoic acid anilide, unii-ak1b12366o, phenyl-n-benzamide, benzanilid, benzoylanilide, n-benzoyl aniline | |
ZVSKZLHKADLHSD-UHFFFAOYSA-N | |
N-phenylbenzamide | |
7168 | |
98% |
Safety and Handling
EINECSNumber : 202-292-7
TSCA : TSCA
RUO â Research Use Only