Learn More
Benzanilide, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1108636
Description
Benzanilide is used to make dyes, active pharmaceutical ingredients and perfumes. It is also used to study the influence of beta-cyclodextrin on photorearrangement of acetanilide, benzanilide and ethyl phenyl carbonate. Further, it is used as an amide model compound to study the reaction between the amide and epoxy. In addition to this, it reacts with aryltriflates in the presence of palladium-based catalyst system to get N-(2,6-diarylbenzoyl)anilines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzanilide is used to make dyes, active pharmaceutical ingredients and perfumes. It is also used to study the influence of beta-cyclodextrin on photorearrangement of acetanilide, benzanilide and ethyl phenyl carbonate. Further, it is used as an amide model compound to study the reaction between the amide and epoxy. In addition to this, it reacts with aryltriflates in the presence of palladium-based catalyst system to get N-(2,6-diarylbenzoyl)anilines.
Solubility
Slightly soluble in diethyl ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Benzanilide | |
162°C to 166°C | |
117°C (10 mmHg) | |
C13H11NO | |
500 g | |
14,1061 | |
Slightly soluble in diethyl ether. Insoluble in water. | |
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2 | |
197.237 | |
197.24 |
93-98-1 | |
1.32 | |
Odorless | |
MFCD00003069 | |
1102980 | |
benzanilide, n-benzoylaniline, benzamide, n-phenyl, n-phenyl-benzamide, benzoic acid anilide, unii-ak1b12366o, phenyl-n-benzamide, benzanilid, benzoylanilide, n-benzoyl aniline | |
ZVSKZLHKADLHSD-UHFFFAOYSA-N | |
N-phenylbenzamide | |
7168 | |
≥98% |
Chemical Identifiers
93-98-1 | |
197.237 | |
ZVSKZLHKADLHSD-UHFFFAOYSA-N | |
7168 | |
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2 |
C13H11NO | |
MFCD00003069 | |
benzanilide, n-benzoylaniline, benzamide, n-phenyl, n-phenyl-benzamide, benzoic acid anilide, unii-ak1b12366o, phenyl-n-benzamide, benzanilid, benzoylanilide, n-benzoyl aniline | |
N-phenylbenzamide |
Safety and Handling
EINECSNumber : 202-292-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only