Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzamidoxime, 95%

$124.45 - $372.18

Chemical Identifiers

Chemical Name or Material Benzamidoxime
CAS 613-92-3
Molecular Formula C7H8N2O
Formula Weight 136.15
MDL Number MFCD00031485
Synonym N-Hydroxybenzenecarboximidamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2606803

  SDS
alfa aesar™
H2606803
1g Each for $124.45

AAH2606806

  SDS
alfa aesar™
H2606806
5g Each for $372.18
Description & Specifications

Specifications

Formula Weight 136.15
Chemical Name or Material Benzamidoxime
Assay 95%
Molecular Formula C7H8N2O
MDL Number MFCD00031485
CAS 613-92-3
Synonym N-Hydroxybenzenecarboximidamide
Health Hazard 1 Warning
Melting Point 72° to 76°C
Quantity 1g
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.