Please login to your online account to display your discounted pricing

Benzamidine Hydrochloride Hydrate (White Crystalline Powder), Fisher BioReagents


Quantity 25g
Packaging Poly Bottle

Manufacturer: fisher scientific company  BP43525

 View more versions of this product

Catalog No. BP435-25

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Powder/Solid
Formula Weight 156.61
Melting Point 167°C
Quantity 25g
Color Off-white
Lambda Max 225 to 235nm
Packaging Poly Bottle
CAS 206752-36-5
Chemical Name or Material Benzamidine Hydrochloride Hydrate
Heavy Metals (as Pb) 0.005% max.
Identification Pass Test
Assay Percent Range ≥99.0 %
Molecular Formula C7H8N2HCl
CAS 1670-14-0
ChemAlert Storage Symbol Gray
Synonym Benzamidinium Chloride
Recommended Storage 4°C

Specifications

Chemical Name or Material Benzamidine Hydrochloride Hydrate
CAS 1670-14-0
CAS 206752-36-5
Physical Form Powder/Solid
Assay ≥99.0%
Assay Percent Range ≥99.0 %
Molecular Formula C7H8N2HCl
Formula Weight 156.61
Synonym Benzamidinium Chloride
Accessories
 

4°C

SDS & Safety Documents