missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzamidine Hydrochloride Hydrate (White Crystalline Powder), Fisher BioReagents

$210.95 - $210.95

Chemical Identifiers

CAS 1670-14-0
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.613
InChI Key LZCZIHQBSCVGRD-UHFFFAOYSA-N
Synonym amidinobenzene hydrochloride
PubChem CID 80289
IUPAC Name benzenecarboximidamide;hydrochloride
SMILES C1=CC=C(C=C1)C(=N)N.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP435-25 SDS View Documents Fisher BioReagents
BP43525
25g Poly Bottle Each for $210.95
Case of 6 Each for $1,265.70
 
Description

Description

Benzamidine Hydrochloride is commonly used as a protease inhibitor.
Specifications

Chemical Identifiers

1670-14-0
156.613
amidinobenzene hydrochloride
benzenecarboximidamide;hydrochloride
C7H9ClN2
LZCZIHQBSCVGRD-UHFFFAOYSA-N
80289
C1=CC=C(C=C1)C(=N)N.Cl

Specifications

Pass Test
White
25g
≥99.0 %
amidinobenzene hydrochloride
C1=CC=C(C=C1)C(=N)N.Cl
156.613
156.61
≥99.0%
Poly Bottle
167°C
1670-14-0,206752-36-5
C7H9ClN2
LZCZIHQBSCVGRD-UHFFFAOYSA-N
benzenecarboximidamide;hydrochloride
80289
Powder/Solid
Benzamidine Hydrochloride Hydrate
Safety and Handling

Safety and Handling

Recommended Storage : 4°C