Learn More
Benzamidine hydrochloride hydrate, 99%, water ca 10-14%, Thermo Scientific Chemicals
Reversible inhibitor of trypsin, trypsin-like proteins and serine proteases
Supplier: Thermo Scientific Chemicals J6282309
Description
Reversible inhibitor of trypsin, trypsin-like proteins and serine proteases
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Benzamidine hydrochloride hydrate | |
80°C to 85°C | |
water ≈10 to 14% | |
MFCD00066285 | |
3594959 | |
LZCZIHQBSCVGRD-UHFFFAOYSA-N | |
benzenecarboximidamide;hydrate;hydrochloride | |
16219042 | |
99% |
206752-36-5 | |
White | |
C7H9ClN2 | |
10 g | |
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate | |
Cl.NC(=N)C1=CC=CC=C1 | |
156.61 | |
156.62 | |
Beads or Powder |
Chemical Identifiers
206752-36-5 | |
156.61 | |
LZCZIHQBSCVGRD-UHFFFAOYSA-N | |
16219042 | |
Cl.NC(=N)C1=CC=CC=C1 |
C7H9ClN2 | |
MFCD00066285 | |
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate | |
benzenecarboximidamide;hydrate;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 216-795-4
RTECSNumber : CV6260000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only