Learn More
Benzamide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 159335000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Benzamide | |
55-21-0 | |
100.0 | |
White | |
180°C | |
98.5% min. (HPLC) | |
C7H7NO | |
MFCD00007968 | |
09, 195 | |
benzoic acid amide, benzoylamide, phenylcarboxyamide, phenylcarboxamide, benzenecarboxamide, amid kyseliny benzoove, amid kyseliny benzoove czech, phenyl carboxyamide, ccris 4594, benzoic acid,amide | |
KXDAEFPNCMNJSK-UHFFFAOYSA-N | |
benzamide | |
2331 | |
121.14 | |
Crystalline Powder |
99% | |
98.5 | |
125.0°C to 129.0°C | |
228.0°C | |
Authentic | |
Plastic bottle | |
C6H5CONH2 | |
500 g | |
15, 1062 | |
Solubility in water: 1g/74mL (20°C). Other solubilities: soluble in hot water,soluble in hot benzene,1g/6mL alcohol- 1g/3.3mL pyridine,slightly soluble in ether | |
C1=CC=C(C=C1)C(=O)N | |
121.14 | |
CHEBI:28179 | |
99% |
Chemical Identifiers
55-21-0 | |
121.14 | |
KXDAEFPNCMNJSK-UHFFFAOYSA-N | |
2331 | |
benzamide |
C7H7NO | |
MFCD00007968 | |
benzoic acid amide, benzoylamide, phenylcarboxyamide, phenylcarboxamide, benzenecarboxamide, amid kyseliny benzoove, amid kyseliny benzoove czech, phenyl carboxyamide, ccris 4594, benzoic acid,amide | |
CHEBI:28179 | |
C1=CC=C(C=C1)C(=O)N |
Safety and Handling
GHS H Statement:
Suspected of causing genetic defects.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-227-7
RTECSNumber : CU8700000
TSCA : TSCA
RUO – Research Use Only