Please login to your online account to display your discounted pricing

Benzamide 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  159330050

 View more versions of this product

Catalog No. AC159330050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 121.14
Quantity 5g
Color White to almost white
Beilstein 09, 195
Merck Index 15, 1062
EINECS Number 200-227-7
TSCA TSCA
Melting Point 125° to 129°C
RTECS Number CU8700000
CAS Max % 100.0
Chemical Name or Material Benzamide
Name Note 99%
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C7H7NO
Linear Formula C6H5CONH2
MDL Number MFCD00007968
CAS 55-21-0
Health Hazard 2 GHS H Statement:
Suspected of causing genetic defects.
Harmful if swallowed.
Infrared Spectrum Authentic
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Benzamide
Name Note 99%
CAS 55-21-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H7NO
Linear Formula C6H5CONH2
Formula Weight 121.14
MDL Number MFCD00007968
Accessories

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing genetic defects.
Harmful if swallowed.

SDS & Safety Documents