Please login to your online account to display your discounted pricing

Benzalphthalide 98%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  105231000

 View more versions of this product

Catalog No. AC105231000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 222.24
Physical Form Crystalline Powder
Quantity 100g
Color Pale yellow to yellow
RTECS Number TI3686000
EINECS Number 209-388-8
Melting Point 100° to 104°C
Beilstein 17, 376
CAS Max % 100.0
Infrared Spectrum Authentic
Synonym 3-Benzylidenephthalide
Name Note 98%
Molecular Formula C15 H10 O2
CAS Min % 97.5
Assay Percent Range 98%
CAS 575-61-1
MDL Number MFCD00005929
Packaging Plastic bottle
Chemical Name or Material Benzalphthalide

Specifications

Chemical Name or Material Benzalphthalide
Name Note 98%
CAS 575-61-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15 H10 O2
Formula Weight 222.24
MDL Number MFCD00005929
Synonym 3-Benzylidenephthalide