Please login to your online account to display your discounted pricing

Benzalphthalide 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Benzalphthalide
Name Note 98%
CAS 575-61-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15 H10 O2
Formula Weight 222.24
MDL Number MFCD00005929
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC105231000

  SDS
thermo fisher scientific
105231000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 222.24
Quantity 100g
Color Pale yellow to yellow
EINECS Number 209-388-8
RTECS Number TI3686000
Melting Point 100° to 104°C
Beilstein 17, 376
CAS Max % 100.0
Infrared Spectrum Authentic
Name Note 98%
Synonym 3-Benzylidenephthalide
Chemical Name or Material Benzalphthalide
Packaging Plastic bottle
Assay Percent Range 98%
Molecular Formula C15 H10 O2
MDL Number MFCD00005929
CAS 575-61-1
CAS Min % 97.5