Please login to your online account to display your discounted pricing

Benzalkonium chloride, alkyl distribution from C8H17 to C16H33, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  215411000

 View more versions of this product

Catalog No. AC215411000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solid or Thick Gel
Color Clear or white to yellow, may contain yellow-white fragments
Quantity 100g
Merck Index 15, 1061
Water 10% Max. (K.F.)
Assay 95.0 to 104.0% (anhydrous basis)
Linear Formula C6H5CH2N(CH3)2RClR=C8H17toC16H33
Synonym Alkyl-benzyl-dimethylammonium chloride
Additional Information Total amines:3.6% max.
Chemical Name or Material Benzalkonium chloride
Packaging Glass bottle
MDL Number MFCD00145757
CAS 8001-54-5
Sulfated Ash 0.5% max.
Infrared Spectrum Authentic
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Do not get in eyes, on skin, or on clothing. Avoid release to the environment. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Get medical attention/advice if you feel unwell.
Health Hazard 2 GHS P Statement Harmful if swallowed. Very toxic to aquatic life. Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material Benzalkonium chloride
Assay 95.0 to 104.0% (anhydrous basis)
Linear Formula C6H5CH2N(CH3)2RClR=C8H17toC16H33
MDL Number MFCD00145757
Synonym Alkyl-benzyl-dimethylammonium chloride
Accessories

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Do not get in eyes, on skin, or on clothing. Avoid release to the environment. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Get medical attention/advice if you feel unwell.

GHS P Statement Harmful if swallowed. Very toxic to aquatic life. Causes severe skin burns and eye damage.

SDS & Safety Documents