Please login to your online account to display your discounted pricing

Benzalkonium chloride, Alfa Aesar™

Cationic surface active agent

Chemical Identifiers

Chemical Name or Material Benzalkonium chloride
CAS 63449-41-2
Molecular Formula C6H5CH2N(CH3)2RCl
MDL Number MFCD00145757
UN Number UN2923
Merck Index 14,1059
Synonym Alkylbenzyldimethylammonium chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA4133936

  SDS
alfa aesar
4133936
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Semi-Solid/Solid, R&Equals;C8H17 To C18H37
Quantity 500g
MDL Number MFCD00145757
Synonym Alkylbenzyldimethylammonium chloride
Molecular Formula C6H5CH2N(CH3)2RCl
Merck Index 14,1059
TSCA TSCA
UN Number UN2923
Solubility Information Soluble in water
CAS 63449-41-2
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H314-H318-H302-H312;
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Chemical Name or Material Benzalkonium chloride
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302-H312;
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.