Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzaldoxime, predominantly (E)-isomer, 98%

$185.34 - $185.34

Chemical Identifiers

Chemical Name or Material Benzaldoxime, predominantly (E)-isomer
CAS 932-90-1
Molecular Formula C7H7NO
Formula Weight 121.14
MDL Number MFCD00002119
Beilstein 774138
Synonym Benzaldehyde oxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1205322

  SDS
alfa aesar™
A1205322
100g Each for $185.34
Description & Specifications

Specifications

Formula Weight 121.14
Molecular Formula C7H7NO
Health Hazard 1 Warning
CAS 932-90-1
Assay 98%
Flash Point 108°C (226°F)
Quantity 100g
Beilstein 774138
Melting Point 33° to 35°C
MDL Number MFCD00002119
Chemical Name or Material Benzaldoxime, predominantly (E)-isomer
Boiling Point 118° to 120°C (10mmHg)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Synonym Benzaldehyde oxime

Applications
Oximes are used as a peel-preventing additive in paints and lacquers. It acts as an antioxidant against oxidative drying materials which forms sticky skin with air oxygen. Oximes are used as chemical building block for the synthesis of agrochemicals and pharmaceuticals. In medicine application, Oxime structure is effective in cholinesterase reactivators to treat the poisoning by organophosphates. Oxime is used as a ligand in transition-metal complex catalyst chemistry. Oxime acts as an antioxidant, radical scavenger which find applications in textile, plastic, paint, detergent, and rubber industry.

Solubility
Slightly soluble in water. Soluble in ethanol and ether.

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases, acids, acid anhydrides and acid chlorides.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.