missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzaldehyde dimethyl acetal, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC209790010

 View more versions of this product

Catalog No. AC209790010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Benzaldehyde dimethyl acetal
69°C
Glass bottle
87.0°C to 89.0°C (18.0mmHg)
1L
1125-88-8
100.0
C9H12O2
MFCD00008491
HEVMDQBCAHEHDY-UHFFFAOYSA-N
dimethoxymethylbenzene
62375
Liquid
1.0100g/mL
Authentic
1.4920 to 1.4940
Colorless
99+%
97.5
98%
C6H5CH(OCH3)2
alpha,alpha-dimethoxytoluene
COC(C1=CC=CC=C1)OC
152.193
152.19
98%

Chemical Identifiers

1125-88-8
152.193
HEVMDQBCAHEHDY-UHFFFAOYSA-N
62375
COC(C1=CC=CC=C1)OC
C9H12O2
MFCD00008491
alpha,alpha-dimethoxytoluene
dimethoxymethylbenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 214-413-0

RTECSNumber : CU5774000

TSCA : TSCA

SDS Documents