missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzaldehyde dimethyl acetal, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC209790025

 View more versions of this product

Catalog No. AC209790025

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Benzaldehyde dimethyl acetal
1125-88-8
100.0
Glass bottle
≥97.5%
1.4920 to 1.4940 (20°C, 589nm)
C6H5CH(OCH3)2
MFCD00008491
Clear colorless
99+%
97.5
Authentic
Liquid
98%
C9H12O2
152.19
1.0100g/mL
2.5L

Chemical Identifiers

1125-88-8
100.0
C9H12O2
152.19
Benzaldehyde dimethyl acetal, 99+%
97.5
98%
C6H5CH(OCH3)2
MFCD00008491
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 214-413-0

RTECSNumber : CU5774000

TSCA : TSCA