Please login to your online account to display your discounted pricing

Benzaldehyde dimethyl acetal, 98%, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: fisher scientific acros  209792500

 View more versions of this product

Catalog No. AC209792500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 152.19
Quantity 250mL
Color Clear colorless
EINECS Number 214-413-0
TSCA TSCA
RTECS Number CU5774000
CAS Max % 100.0
Chemical Name or Material Benzaldehyde dimethyl acetal
Name Note 99+%
Assay Percent Range 98%
Molecular Formula C9H12O2
Linear Formula C6H5CH(OCH3)2
MDL Number MFCD00008491
CAS 1125-88-8
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4920 to 1.4940 (20°C, 589nm)
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Benzaldehyde dimethyl acetal
Name Note 99+%
CAS 1125-88-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H12O2
Linear Formula C6H5CH(OCH3)2
Formula Weight 152.19
MDL Number MFCD00008491
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents