Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzaldehyde azine, 99%

$52.34 - $546.45

Chemical Identifiers

Chemical Name or Material Benzaldehyde azine
CAS 588-68-1
Molecular Formula C14H12N2
Formula Weight 208.27
MDL Number MFCD00016876
Beilstein 2208840
Synonym Benzalazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1572114

 
alfa aesar™
A1572114
25g Each for $52.34

AAA1572122

 
alfa aesar™
A1572122
100g Each for $136.85

AAA1572136

 
alfa aesar™
A1572136
500g Each for $546.45
Description & Specifications

Specifications

Formula Weight 208.27
Synonym Benzalazine
Chemical Name or Material Benzaldehyde azine
TSCA TSCA
Assay 99%
Molecular Formula C14H12N2
MDL Number MFCD00016876
CAS 588-68-1
Beilstein 2208840
Health Hazard 1 Warning
Melting Point 91° to 95°C
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number CU7571500
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.