missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzaldehyde azine, 99%

Manufacturer:  Alfa Aesar™ A1572114

 View more versions of this product

Catalog No. AAA1572114



Specifications

Specifications

Benzaldehyde azine
25g
C14H12N2
2208840
CWLGEPSKQDNHIO-NFLUSIDLSA-N
(Z)-N-[(Z)-benzylideneamino]-1-phenylmethanimine
5371793
99%
91°C to 95°C
588-68-1
MFCD00016876
1,2-dibenzylidenehydrazine, n-z-benzylideneamino-1-phenylmethanimine, 1,2-di e-benzylidene hydrazine, e-n-e-benzylideneamino-1-phenylmethanimine
C1=CC=C(C=C1)C=NN=CC2=CC=CC=C2
208.264
208.27

Chemical Identifiers

588-68-1
208.264
CWLGEPSKQDNHIO-NFLUSIDLSA-N
5371793
C1=CC=C(C=C1)C=NN=CC2=CC=CC=C2
C14H12N2
MFCD00016876
1,2-dibenzylidenehydrazine, n-z-benzylideneamino-1-phenylmethanimine, 1,2-di e-benzylidene hydrazine, e-n-e-benzylideneamino-1-phenylmethanimine
(Z)-N-[(Z)-benzylideneamino]-1-phenylmethanimine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CU7571500

TSCA : TSCA

Documents