missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzal Chloride, TCI America™

$39.72 - $256.67

Chemical Identifiers

CAS 98-87-3
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000829
InChI Key CAHQGWAXKLQREW-UHFFFAOYSA-N
Synonym benzal chloride, dichloromethyl benzene, alpha,alpha-dichlorotoluene, benzylidene chloride, benzene, dichloromethyl, benzyl dichloride, benzylene chloride, chlorobenzal, dichlorophenylmethane, chlorure de benzylidene
PubChem CID 7411
ChEBI CHEBI:82273
IUPAC Name dichloromethylbenzene
SMILES C1=CC=C(C=C1)C(Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B141425G View Documents TCI America
B141425G
25g Each for $39.72
 
B1414500G View Documents TCI America
B1414500G
500g Each for $256.67
 
B1414100G View Documents TCI America
B1414100G
100g Each for $100.83
 
Specifications

Chemical Identifiers

98-87-3
161.025
CAHQGWAXKLQREW-UHFFFAOYSA-N
7411
dichloromethylbenzene
C7H6Cl2
MFCD00000829
benzal chloride, dichloromethyl benzene, alpha,alpha-dichlorotoluene, benzylidene chloride, benzene, dichloromethyl, benzyl dichloride, benzylene chloride, chlorobenzal, dichlorophenylmethane, chlorure de benzylidene
CHEBI:82273
C1=CC=C(C=C1)C(Cl)Cl

Specifications

207°C
-16°C
98-87-3
MFCD00000829
benzal chloride, dichloromethyl benzene, alpha,alpha-dichlorotoluene, benzylidene chloride, benzene, dichloromethyl, benzyl dichloride, benzylene chloride, chlorobenzal, dichlorophenylmethane, chlorure de benzylidene
C1=CC=C(C=C1)C(Cl)Cl
161.025
CHEBI:82273
Clear Liquid at 20°C
Benzal Chloride
Yellow
25g
C7H6Cl2
1886
CAHQGWAXKLQREW-UHFFFAOYSA-N
dichloromethylbenzene
7411
161.03
≥95.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0078&(3)-0101

RTECSNumber : CZ5075000

TSCA : Yes