Please login to your online account to display your discounted pricing

Alfa Aesar™ Benz[a]anthracene-7,12-dione, 97%

$257.92 - $257.92

Chemical Identifiers

Chemical Name or Material Benz[a]anthracene-7,12-dione
CAS 2498-66-0
Molecular Formula C18H10O2
Formula Weight 258.28g/mol
MDL Number MFCD00003596
Beilstein 2052941
Synonym 1,2-Benzanthraquinone; C.I. 59000
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1659314

 
alfa aesar™
A16593-14
25g Each for $257.92
Description & Specifications

Specifications

Melting Point 167° to 172°C
Quantity 25g
Chemical Name or Material Benz[a]anthracene-7,12-dione
CAS 2498-66-0
Assay 97%
Molecular Formula C18H10O2
Formula Weight 258.28g/mol
MDL Number MFCD00003596
Beilstein 2052941
Synonym 1,2-Benzanthraquinone; C.I. 59000
RTECS Number CW4591000
TSCA TSCA