Please login to your online account to display your discounted pricing

Benidipine hydrochloride, Tocris™

Calcium channel blocker

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material Benidipine hydrochloride
Assay Percent Range >99%
Molecular Formula C28H31N3O6·HCl
Formula Weight 542.02
Synonym (4R)-rel-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 3-methyl 5-[(3R)-1-(phenylmethyl)-3-piperidinyl] ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

393410

 
r & d systems
RDS393410
10mg Each for $201.36

393450

 
r & d systems
RDS393450
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 542.02
Quantity 10mg
Recommended Storage Desiccate at Room Temperature
Assay Percent Range >99%
Chemical Name or Material Benidipine hydrochloride
Synonym (4R)-rel-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 3-methyl 5-[(3R)-1-(phenylmethyl)-3-piperidinyl] ester hydrochloride
CAS 91599-74-5
Molecular Formula C28H31N3O6·HCl
Safety and Handling

Desiccate at Room Temperature