Benedict Quantitative Solution, Certified, 1.95 to 2.05mg/mL equivalence, LabChem™
Supplier: LabChem LC116601
Specifications
Benedict Quantitative Solution | |
58.21,20.04,12.52,7.41,1.8 | |
CuSO4·5H2O | |
Soluble in water | |
Certified | |
Poly Bottle | |
500 mL |
Blue | |
63126-89-6,333-20-0,497-19-8,7758-99-8 | |
1.95 to 2.05mg/mL equivalence | |
249.68 | |
Passes Test | |
Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Sulfur compounds; Copper | |
Liquid |