missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Bench Mounted Reagent Poles

Fisherbrand™ Bench Mounted Reagent Poles are used to make stackable shelving on an island or peninsula. Poles can be stacked to make multiple tier shelving.

Manufacturer:  Fisherbrand™ FRP15

 View more versions of this product

Catalog No. 06000109



Includes: Hardware

Description

Description

  • 1 in. Diameter painted steel pole is threaded at both ends
  • Bolts through the work surface for secure mounting
  • Fastener on the reagent shelf surface is recessed
  • Hardware is included for each installation
  • Poles are painted Fisherbrand White
Specifications

Specifications

White
1 in.
38.1cm
Steel
1 in.
2.5cm
15 in.
Installation Hardware
Reagent Pole
2.5cm
Documents