missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Wilmad™ Bottom Plug for 3.2mm MAS Rotor

Designed for use with MAS Rotors. Bel-Art SP Scienceware™ Wilmad™ Bottom Plug for 3.2mm MAS Rotor features a Vespal composition and works with applications ranging in temperature from -30° to 70°:C.

Supplier:  Bel-Art™ 660000103

Catalog No. 16-800-203



Description

Description

  • For 3.2mm Bruker™ MAS Probe
Specifications

Specifications

3.2 mm Bruker MAS Probe:
Vespel
-30°C to +70°C
Bottom Plug
WP-602-3182
SDS
Documents

Documents

Promotions

Promotions