Bel-Art™ SP Scienceware™ Wire Lead Flask with Vikem™ Coating
“Wire” lead weights coil into a helix, spiral, cone or any irregular shape to stabilize vessels in a water bath
Manufacturer:  Bel-Art™ F183240001
 View more versions of this product
Catalog No. 15-547
Description
Description
  • Can be coiled around flasks, threaded through handles or looped around bottle necks
  • Coated with chemical resistant red Vikem vinyl
  • The 30cm (12 in.) length and flexibility make these weights very versatile
Specifications
Specifications
No
0.8cm
0.38 lb.
Accessory
0.31 in.
1
176g
Documents