Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Vacuum Aspirator Bottles
Offers easy and continuous aseptic vacuum aspiration of liquids, including tissue culture media, supernatants and chemical wastes
$620.74 - $777.32
Specifications
Autoclavable Yes
Material Polypropylene
For Use With (Application) For dishes, plates, flasks and tubes
View More Specs
Products
Catalog Number Mfr. No. Holds Capacity (English) Capacity (Metric) Price Quantity    
13-717-25
bel-art™
F199170002
3200mL and the included trap prevents fluid flow into the vacuum source 1 gal. 3.78L Each for $777.32
13-717-24
bel-art™
199170001
1600mL and the included trap prevents fluid flow into the vacuum source 0.5 gal. 1.89L Each for $620.74
Description & Specifications
Specifications
Autoclavable Yes
Material Polypropylene
For Use With (Application) For dishes, plates, flasks and tubes
  • When used with a vacuum source, allows aspiration from a pipet tip, Pasteur or serological pipet inserted into the collection tube
  • Chemical and corrosion-resistant polypropylene bottle and epoxy-coated steel frame
  • Compact design does not crowd workspace or fume hood
  • Included trap prevents fluid flow into the vacuum source
  • Suitable for use with dishes, plates, flasks and tubes
  • Bottle and cap are autoclavable at 121°C (250°F)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.