Bel-Art™ SP Scienceware™ Urbanti™ High-Speed Filter Funnels
Helicoid internal ribs greatly increase filtration speed.
$40.80 - $46.00
Specifications
Autoclavable Yes
Diameter (English) Outer Stem 0.39 in.
Material Polymethylpentene
Includes Internal helicoid ribs
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (English) Diameter (English) Top Length (English) Stem Price Quantity  
Catalog Number Mfr. No. Capacity (English) Diameter (English) Top Length (English) Stem Price Quantity  
S34514-1 Bel-Art™
F14644EMD
2.56 oz. 2.75 in. 2.75 in. Pack of 6 Each for $46.00
S345131 Bel-Art™
F14640EMD
0.95 oz. 2 in. 5.9 in. Pack of 6 Each for $40.80
S34515 Bel-Art™
F146460000EMD
7.57 oz. 3.93 in. 4.2 in. Pack of 4 Each for $41.70
Specifications
Description
  • Molded polymethylpentene funnel
  • Uses standard, round filter papers
  • Made to withstand corrosive or volatile liquids and temperatures up to 177°C (350°F)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Yes
0.39 in.
Polymethylpentene
Internal helicoid ribs