missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Two-Color Wash Bottles - Safety-Vented™ and Safety Labeled, Wide-Mouth
A unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know Safety-Labeled Wash Bottles
Manufacturer: Bel-Art™ F116430223
Alert:
Use only with solvents compatible with LDPE.
Description
A unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know Safety-Labeled Wash Bottles
Volatiles are vented eliminating solvent drips. They are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.
- Valve eliminates drips; allows dispensing of wash bottle in an upright or inverted position
- Wide mouth 53mm (2 in.) bottle for easy refilling
- Polypropylene closure with leakproof design; color-coded to aid in easy identification
- Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
- Labeled with six of the most commonly used solvent names or label your own; custom labeling available
Order Info
8 oz. (250mL)
Specifications
Not Autoclavable | |
53 mm | |
8 oz., 250mL | |
8 oz. | |
Green | |
Wash Bottle | |
Green |
Polypropylene | |
Wide | |
250 mL | |
Methanol | |
Methanol | |
LDPE |