Bel-Art™ SP Scienceware™ TPX™ Graduated Pitchers
Offer well-designed spouts and large handles helpful for use with wet or dry ingredients
$31.66 - $68.86
Specifications
Material PMP (Polymethylpentene)
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
22-261461 Bel-Art™
F289840000
2000mL Each for $68.86
Case of 12 Each for $826.32
03-410-807 Bel-Art™
F289820000
500mL Each for $31.66
Case of 24 Each for $759.84
03-410-808 Bel-Art™
F289830000
1000mL Each for $44.14
Case of 24 Each for $1,059.36
Specifications
Description
  • Crystal clear polymethylpentene pitcher shows metric graduations molded on the outside wall
  • Chemical resistant
  • Shatterproof
  • Heat resistant to 177°C (350°F)
  • Steam autoclavable at 121°C (215°F)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
PMP (Polymethylpentene)