missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ TPX™ Graduated Pitchers

Offer well-designed spouts and large handles helpful for use with wet or dry ingredients

Manufacturer:  Bel-Art™ F289820000

 View more versions of this product

Catalog No. 03-410-807




Description

Description

  • Crystal clear polymethylpentene pitcher shows metric graduations molded on the outside wall
  • Chemical resistant
  • Shatterproof
  • Heat resistant to 177°C (350°F)
  • Steam autoclavable at 121°C (215°F)

Specifications

Specifications

Yes
Pitcher
500mL
PMP (Polymethylpentene)
Documents