Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Touch Free™ Automatic Waste Cans


Automatically open and close without a touch, protecting users from waste receptacle contact

Manufacturer: bel-art™  132020030

 View more versions of this product

Requires four “D” batteries (not included) which will provide approximately 4000 openings. All cans should be lined with disposable trash or biohazard bags (sold separately). Batteries and bags available on www.fishersci.com.

Catalog No. 11-394-453

  • / Each
 

Description & Specifications

Specifications

Length (Metric) 36.8cm
Width (Metric) 27cm
Capacity 12 gal., 45.5L
Height (Metric) 67.3cm
Capacity (Metric) 45.5L
Height (English) 26.5 in.
Material Stainless Steel
Dimensions (L x W x H) 14.5 x 10.5 x 26.5 in. (36.8 x 27 x 67.3cm)
Product Type Automatic Waste Can
Width (English) 10.5 in.
Length (English) 14.5 in.
Color Black Top
Capacity (English) 12 gal.
Includes Brightly colored biohazard label and three other labels (recyclables, trash, and a blank label for individual applications)