missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Techni-Dome™ 360 Fume Hood
SP Scienceware™ Techni-Dome™ 360 Fume Hood is designed for micropipetting protocols, mixing of reagents, and manipulating small biohazardous items or as a technical demonstration chamber.
Manufacturer: Bel-Art™ F500000100
Description
Offers generous interior workspace with a small footprint
- Compact; fits on crowded benchtops
- Clear, unrestricted viewing from all angles
- Polycarbonate plastic; resistant to a wide range of chemicals
- Smooth interior for easy cleaning
- High impact strength and scratch-resistance
- Built-in handles makes it portable and suited for field use while both halves of the unit nest for compact storage
- Stepped outlet connects to standard exhaust systems with 4 or 6 in. vent piping
- Overall volume, 65L (4000 cu. in.)
- Overall dimensions 60W x 60cm H (22W x 22H in.)
Specifications
Fume Hood | |
Polycarbonate | |
5kg |
Polycarbonate | |
11 lb. |