Bel-Art™ Scienceware™ Switch-Grid™ Test Tube Racks
One rack provides 25 possible tube size combinations
Manufacturer:  Bel-Art™ F187450011
 View more versions of this product
Catalog No. 11-123-988
Specifications Description & Specifications
Specifications
Rack with Blue Grids
12.5 x 4.37 x 2.62 in. (31.8 x 11.1 x 6.7cm)
6.7cm
12.5 in.
Plastic
Rows: 12x5
11.1cm
Blue
2.62 in.
13 to 16mm Tubes
31.8cm
60
4.37 in.
Accessories