missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Super Biohazard Disposal Bags
Protects against dangerous tears and leaks. SP Scienceware™ Super Biohazard Disposal Bags feature a sterilization indicator patch that darkens when exposed to steam sterilization for 20 minutes.
Manufacturer: Bel-Art™ F131651923
Description
- Protect against dangerous tears and leaks
- Orange-red bags have standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish
- Extra-heavy-duty bags are 2mil-thick blended polymer
- Autoclavable to 121°C (250°F)
- Sterilization indicator patch darkens when exposed to steam sterilization
Specifications
23 x 19 in. (58 x 48 cm) | |
48 cm | |
Orange Red | |
Standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish | |
Polymer | |
Biohazard Disposal Bag |
58 cm | |
Autoclavable | |
Extra-strength, high-impact, high-temperature bags reduce autoclave time | |
23 in. | |
2 mil | |
19 in. |