missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Super Biohazard Disposal Bags
Protects against dangerous tears and leaks. SP Scienceware™ Super Biohazard Disposal Bags feature a sterilization indicator patch that darkens when exposed to steam sterilization for 20 minutes.
$587.44 - $856.41
Specifications
Autoclavable | Autoclavable |
---|---|
Color | Orange Red |
For Use With (Application) | Extra-strength, high-impact, high-temperature bags reduce autoclave time |
Includes | Standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish |
Material | Polymer |
Description
- Protect against dangerous tears and leaks
- Orange-red bags have standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
- Extra-heavy-duty bags are 2 mil-thick blended polymer
- Autoclavable to 121°C (250°F)
- Sterilization indicator patch darkens when exposed to steam sterilization
Specifications
Autoclavable | |
Extra-strength, high-impact, high-temperature bags reduce autoclave time | |
Polymer | |
2 mil |
Orange Red | |
Standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish | |
Biohazard Disposal Bag |