missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Stir and Add™ Cuvet Mixer

Made of clear acrylic with a straight handle, this tool is used when reactions are performed directly into cuvettes, to mix added reagents

$876.07 - $876.07

Specifications

Diameter (English) Shaft 0.125 in.
Diameter (Metric) Shaft 3.17mm
Case Quantity 36 Case
Material Acrylic
Packaging Bag
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Cell Type Length (English) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Cell Type Length (English) Length (Metric) Price Quantity  
14-511-20 Bel-Art™
F377520000
10mm light path 4.5 in. 11.4cm Case of 36 Each for $876.07
14-511-21 Bel-Art™
F377520001
Semi-micro 5.1 in. 13cm Case of 36 Each for $876.07
Specifications

Description

  • Up and down motion in cell provides complete mixing
  • Shaft diameter 3.17mm (0.125 in.)
  • Straight handle

Used to add enzymes and other materials to solution in spectrophotometer cells.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

0.125 in.
36 Case
Bag
Straight
3.17mm
Acrylic
Used to add enzymes and other materials to solution in spectrophotometer cells, this tool is used when reactions are performed directly into cuvettes, to mix added reagents