Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Sterilizing Tray Covers
For use with sterilizing trays.
$406.29 - $583.47
Specifications
Material Polypropylene
Description Tray cover
View More Specs
Products
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity    
22-087270
bel-art™
H162590000
Fits 5L tray, 30.5L x 20.3W x 12.7cm H Each for $406.29
22-087-395
bel-art™
162630000
Fits 18L tray, 53.3L x 39.4W x 12.7cm H Each for $583.47
Description & Specifications
Specifications
Material Polypropylene
Description Tray cover
  • Polypropylene
  • Fits snugly over rim of tray
  • Autoclavable at 121°C (250°F)