Bel-Art™ SP Scienceware™ Sterileware™ Sampling Spatulas
Ideal for every type of powdered or granulated material. Bel Art™ SP Scienceware™ Sterileware™ Sampling Spatulas are perfect for taking sterile samples during process quality inspections and for piercing plastic-lined paper bags or similar packages.
$60.33 - $405.76
Specifications
Product Type Sterileware™ Sampling Spatula
Disposable Yes
Material Polystyrene
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Length (English) Sterile Length (Metric) Price Quantity  
Catalog Number Mfr. No. Includes Length (English) Sterile Length (Metric) Price Quantity  
03-421-149 Bel-Art™
F378400000
6 in. L V-shaped channel 9 in. No 22.9cm Dozen of 12 Each for $60.33
Case of 12 Dozen for $723.96
14-358-20 Bel-Art™
H369300000
6 in. L V-shaped channel 9 in. Yes 22.9cm Case of 100 Each for $405.76
16-241-210 Bel-Art™
H369300050
10 in. L V-shaped channel 14 in. Yes 35.6cm Case of 50 Each for $244.88
Specifications
Description
Time-honored V-shape makes transferring powdered, flaked, crystalline or granular media a breeze
Long, 14 in. spatula (16-241-210) is perfect for dislodging caked or crystallized reagents from deep within tall bottles or vessels having a neck diameter as small as 0.75 in. (18mm)
Sterileware (16-241-210, 14-385-20)
  • 9 and 14 in. models
  • High-impact polystyrene
  • Individually packaged
  • Sterilized by gamma irradiation
  • Lot stamped, shipped with Certificate of Processing
Nonsterile Model (03-421-149)
  • 9 in. spatula
  • Polypropylene
Description & Specifications
Specifications
Sterileware™ Sampling Spatula
Yes
Polystyrene