missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Spinplus™ Magnetic Stirring Bars

Spinplus™ magnetic stirring bars add speed and efficiency to mixing operations

Manufacturer:  Bel-Art™ F371440034

 View more versions of this product

Catalog No. S50399-2




Description

Description

  • The “+” shape creates a deep vortex and provides stable, quiet operation
  • Alnico magnet encapsulated in FDA grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe

Specifications

Specifications

19mm
50
Yes
White
0.75 in.
Alnico Magnet
Cross
FDA, USP Class VI, ISO 9001:2008
0.75 in.
19mm
SDS
Documents
Product Certifications
Promotions

Promotions