missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Spinfin™ Magnetic Stirring Bars

Agitate liquids in round bottom vessels like test tubes, cylinders and flasks

Manufacturer:  Bel-Art™ F371250012

 View more versions of this product

Catalog No. 14-512-168




Description

Description

  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating
  • Does not wobble in vessel
  • Autoclave safe

Certifications

ISO 9001:2008, SGS certification.

Specifications

Specifications

Yes
PTFE
0.5 in.
12.7mm
Stir bar
Alnico Magnet
FDA, USP Class VI, ISO 9001:2008, SGS
White
0.5 in.
50
12.7mm
Documents