missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Spinbar™ Octagon Rare Earth Magnetic Stir Bar
Rare Earth/Samarium Cobalt magnet provides strong coupling with drive magnets to reduce frequency of spinout
Supplier: Bel-Art™ F371020138
Description
- Well-suited for viscous solutions, high speed stirring and extended stirring applications
- Octagon shape with pivot ring creates strong turbulance with minimal chattering and friction
- Coated with Teflon™ PTFE
- Distinctive green color for easy identification and visibility
- Autoclave safe
Specifications
9.5 mm | |
0.37 in. | |
1 | |
Autoclavable | |
25.4 mm |
1 in. | |
Green | |
Samarium Cobalt Magnet | |
FDA, ISO 9001:2008, SGS | |
Octagon |