Bel-Art™ SP Scienceware™ SMORC™ Small Orifice Cleaner
Uniquely designed for cleaning and unclogging small orfices; a pin vise securely holds a stainless steel cleaning wire in place
$151.91 - $237.63
Specifications
For Use With (Application) Clean and clear small opening such as sample and reagent probe, reusable syringes, tubing
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Equipment) Length (English) Length (Metric) Product Type Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Length (English) Length (Metric) Product Type Price Quantity  
13-717-12 Bel-Art™
H381260000
Small orifice cleaner 2 in. 5.1cm Small orifice cleaner replacement wire Pack of 30 Each for $151.91
11-394-302 Bel-Art™
F381250000
Probe, reusable syringe, tubing 4 in. 10.2cm Small orifice cleaner Each for $237.63
Case of 12 Each for $2,851.56
Specifications
Description
  • 2.2cm (7/8 in.) diameter ring magnet on the handle is useful for picking up wires and for preventing the unit from rolling on metal surfaces
  • Length without wire is 10.2cm (4 in.)
  • Includes H38126-0000 wire assortment; stores conveniently inside the hollow handle
  • Wire assortment contains 30 stainless steel wires, 5.1cm (2 in.) long, packaged in a plastic vial
  • 10 each of 3 sizes: 0.152, 0.254, 0.508mm (0.006, 0.010, 0.020 in.)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Clean and clear small opening such as sample and reagent probe, reusable syringes, tubing