missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Slide Staining and Storage Apparatus
Conveniently stain and store gels
$62.21 - $178.86
Specifications
Color | Black |
---|---|
For Use With (Application) | Used in cytology and histology labs |
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
22-261-921
|
Bel-Art™
F441820000 |
Staining Dish |
Each for $178.86
Case of 24 Each for $4,292.64 |
|
22-261-920
|
Bel-Art™
F441800000 |
Slide Rack |
Each for $62.21
Case of 24 Each for $1,493.04 |
|
Description
- Versatile slide rack and vessel for radiography gel staining and staining protocols
- PMPA plastic construction makes the unit virtually shatterproof, lightweight, with exceptional solvent and heat resistance for extended use in cytology and histology labs
- The rack and dish can simply be cleaned and are highly resistant to standard stains used in staining protocols
Specifications
Black | |
Used in cytology and histology labs |