Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Vertical Models


Constructed of Durastar® co-polyester. Bel Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing and chemical attack.

Manufacturer: bel-art™  F420741007

 View more versions of this product

Catalog No. 08-647-676

  • / Each
 

Description & Specifications

Specifications

Model 4.0
Includes Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
Width (English) Door 8.9 in.
Depth (English) 16.3 in.
Cabinet Style Vertical
Door Style Left-hinged and gasketed
Height (Metric) Exterior 51.8cm
Autoclavable No
Shelves 3
Shelf Type Removable perforated shelves
Depth (Metric) 41.4cm
Volume (English) 1.9 cu. ft.
Height (English) Opening 16.4 in.
Material Copolyester
Width (Metric) 34.1cm
Width (Metric) Door 22.6cm
For Use With (Application) Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250
Resistance Chemicals, stains and fine cracks
Color Blue with Clear Door
Width (English) 13.4 in.
Height (English) Exterior 20.4 in.

  • Amber color models reduce visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
  • Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
  • Compact 34.1W x 41.4cmD footprint for space saving stacking up to 3 units high
  • Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
  • Can be used with desiccant cartridges, F420450250, F420450200, F420480100