Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Vertical Models


Constructed of Durastar® co-polyester. Bel Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing and chemical attack.

Manufacturer: bel-art™  F420731000

 View more versions of this product

Catalog No. 08-647-657

  • / Each
 

Description & Specifications

Specifications

Width (Metric) 34.1cm
Volume (English) 1.58 cu. ft.
Material Copolyester
Width (English) 13.4 in.
Resistance Chemicals, stains and fine cracks
Length (Metric) 41.4cm
Height (Metric) Exterior 41.7cm
Height (English) Exterior 16.4 in.
Autoclavable No
Dimensions (L x W) 16.3 x 13.4 in. (41.4 x 34.1cm)
Shelf Type Removable perforated shelves
Includes Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
Depth (English) 16.3 in.
Width (English) Door 8.9 in.
Height (English) Opening 12.3 in.
Depth (Metric) 41.4cm
Color Clear
Door Style Left-hinged and gasketed
Model 3.0
For Use With (Application) Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250
Width (Metric) Door 22.6cm
Length (English) 16.3 in.
Cabinet Style Vertical
Shelves 3

  • Amber color models reduce visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
  • Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
  • Compact 34.1W x 41.4cmD footprint for space saving stacking up to 3 units high
  • Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
  • Can be used with desiccant cartridges, F420450250, F420450200, F420480100