Bel-Art™ SP Scienceware™ Secador™ Auto Desiccator Cabinets: Vertical Models
A UV light blocking, dry environment with automatic electronic dehumidification
Manufacturer:  Bel-Art™ F420741106
 View more versions of this product
Catalog No. 08-647-678
Specifications Description & Specifications
Specifications
1.87 cu. ft.
5° to 40°C
3
20.4 in.
16.3 x 13.4 in. (41.4 x 34.1cm)
16.3 in.
41.4cm
41.4cm
Chemicals, stains and fine cracks
Copolyester
Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant
51.8cm
4.0
13.4 in.
Vertical
Perforated removable shelves
Clear with Blue End Caps
Side-swinging
16.3 in.
Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
41° to 104°F
100V
34.1cm
No