Bel-Art™ SP Scienceware™ Replacement Washers for the Pipette Pump™ Pipettors
Use with SP Scienceware™ Pipette Pump™ pipettors. Bel-Art™ SP Scienceware™ Replacement Washers are molded silicone rubber replacement parts. 3/Pack.
Supplier: Bel-Art™ H937890081
Description
- For use with Pipette Pump pipettor with thumbwheel and plunger aspiration
- Replacement washers to fit various sizes/colors of Pipette Pumps in case of loss or damage
- Molded silicone rubber is resistant to acids and alkalis
- Disassemble parts for cleaning and before autoclaving at 121°C (250°F) for 20 minutes
Specifications
Not Autoclavable | |
Washers | |
10mL pipette pump pipettor |