Bel-Art™ SP Scienceware™ Reducing Quick-Disconnects
Each consists of two barbed polyethylene connectors which assemble tightly together with a male-female center taper
Manufacturer:  Bel-Art™ H197290000
 View more versions of this product
Catalog No. 15-315-6C
Description
Description
Interchangeable male/female polyethylene connections
  • All sizes are interchangeable with others to make reducing combinations
  • For use in connecting and disconnecting vacuum lines and other tubing assemblies
  • Operate at temperatures up to 80°C (176°F)
Specifications
Specifications
0.375 to 0.5 in.
Quick DisConnect
Two barbed connectors, one male and one female
Polyethylene
9 to 12mm
Documents