Bel-Art™ SP Scienceware™ Reducing Quick-Disconnects
Each consists of two barbed polyethylene connectors which assemble tightly together with a male-female center taper
Supplier: Bel-Art™ H197280000
Description
Interchangeable male/female polyethylene connections
- All sizes are interchangeable with others to make reducing combinations
- For use in connecting and disconnecting vacuum lines and other tubing assemblies
- Operate at temperatures up to 80°C (176°F)
Specifications
Quick DisConnect | |
Polyethylene | |
Two barbed connectors, one male and one female |
0.25 to 0.375 in. | |
6 to 8mm |