Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Quaver™ and Quaverette™ Vibrating Spatulas
Accurately transfer small amounts of material
$107.79 - $158.53
Specifications
Blade Material Stainless Steel
For Use With (Equipment) Weighing dish or weighing boat or container
View More Specs
Products
Catalog Number Mfr. No. Coating Includes Length (Metric) Price Quantity    
22-385039
bel-art™
H367140000
N/A Angled end, toothed thumb wheel 18cm Each for $107.79
Case of 24 Each for $2,586.96
14-430
bel-art™
367180000
N/A Toothed thumb wheel 20.95cm Each for $158.53
Case of 24 Each for $3,804.72
Description & Specifications
Specifications
Blade Material Stainless Steel
For Use With (Equipment) Weighing dish or weighing boat or container
  • Rotating the toothed thumb wheel vibrates the spatula, giving user control over transfer from spatula to weighing dish
  • Make accurate additions of precious materials as needed
  • Quaverette Micro-Blade Vibrating Spatula permits utilization of even smaller quantities and containers; Narrow V-shaped blade maneuvers easily into small mouth containers and tight corners
  • Fluo-Kem™ Teflon™ FEP coating complies with FDA regulations governing direct food contact applications
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.